missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS Antibody (669238), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB69752JF549
400.35 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
ICOS Monoclonal antibody specifically detects ICOS in Human samples. It is validated for Flow Cytometry
Specifications
| ICOS | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
| Partial recombinant human ICOS protein (amino acids 21-141) [UniProt Q9Y6W8] | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 29851 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 669238 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction