missing translation for 'onlineSavingsMsg'
Learn More

Icomethasone, TRC

Product Code. 30402479
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402479 50 mg -
30393424 500 mg 500mg
30403669 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402479 Supplier Toronto Research Chemicals Supplier No. TRCI16410050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4647-20-5
Molecular Formula C22 H29 Cl O5
Molecular Weight (g/mol) 408.92
Synonym Pregna-1,4-diene-3,20-dione, 9-chloro-11,17,21-trihydroxy-16-methyl-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 9-chloro-11β,17,21-trihydroxy-16α-methyl- (7CI,8CI), (11β,16α)-9-Chloro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, Icometasone, Icomethasone, Mometasone Furoate Imp. K (EP)
IUPAC Name (8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H29ClO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1
Formula Weight 408.1704
Chemical Name or Material Icomethasone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.