missing translation for 'onlineSavingsMsg'
Learn More

Iclaprim, MedChemExpress

Product Code. 16453388
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16453388 10 mg 10mg
16439916 50 mg 50mg
16430769 1 mg 1mg
16424626 100 mg 100mg
16416117 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16453388 Supplier Medchem Express Supplier No. HY101479
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Iclaprim is a new selective bacterial Dihydrofolate inhibitor, which can inhibit the growth of S. aureus (MRSA) with an MIC90 of 0.06 μg/mL.

Iclaprim is a new selective bacterial Dihydrofolate inhibitor, which can inhibit the growth of S. aureus (MRSA) with an MIC90 of 0.06 μg/mL.

Specifications

Chemical Name or Material Iclaprim
CAS 192314-93-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H22N4O3
Quantity 10 mg
Synonym AR-100
Solubility Information DMSO : 30 mg/mL (84.65 mM; Need ultrasonic and warming)
SMILES NC1=NC=C(CC2=C3C=CC(C4CC4)OC3=C(OC)C(OC)=C2)C(N)=N1
Molecular Weight (g/mol) 354.4
Formula Weight 354.4
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.