missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551513
This item is not returnable.
View return policy
Description
Immunogen sequence: VGHPLGSTTQ NLQDSEKPQK GILEKAGPPP YIKPVPPAQP PAKPHTRISS RQHQASQPPL HLTYPYKAEV SRTDHPSHLQ EDKPSPLLFP SLHNKHPQSK ITAGLEHKNG EIKPKSRRR Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 63%.
ICK or intestinal cell kinase belongs to a very extensive family of proteins that share a conserved catalytic core common with both serine/threonine and tyrosine protein kinases. ICK encodes an intestinal serine/threonine kinase harboring a dual phosphorylation site found in mitogen-activating protein (MAP) kinases. ICK localizes to the intestinal crypt region and is thought to be important in intestinal epithelial cell proliferation and differentiation. ICK purified from transfected human embryonic kidney cells undergoes autophosphorylation and phosphorylates a test protein. ICK plays a key role in development of central nervous, skeletal, and endocrine systems.
Specifications
| ICK | |
| Polyclonal | |
| Unconjugated | |
| CILK1 | |
| 2210420N10Rik; AI848300; ciliogenesis associated kinase 1; CILK1; ECO; heart serine-threonine protein kinase; hICK; iciliogenesis associated kinase 1; ICK; intestinal cell (MAK-like) kinase; intestinal cell kinase; KIAA0936; Laryngeal cancer kinase 2; LCK2; MAK-related kinase; mICK; MRK; serine/threonine protein kinase; serine/threonine-protein kinase ICK | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22858 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UPZ9 | |
| CILK1 | |
| Recombinant protein corresponding to Human ICK. Recombinant protein control fragment (Product #RP-93677). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction