missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68573-25ul
This item is not returnable.
View return policy
Description
ICK Polyclonal antibody specifically detects ICK in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| ICK | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| EC 2.7.11, hICK, intestinal cell (MAK-like) kinase, Intestinal cell kinase, KIAA0936EC 2.7.11.22, Laryngeal cancer kinase 2, LCK2serine/threonine protein kinase, MAK-related kinase, MGC46090, MRKECO, serine/threonine-protein kinase ICK | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DFSPSLSRIDLKNKKRQSDDTLCRFESVLDLKPSEPVGTGNSAPTQTSYQRRDTPTLRSAAKQHYLKHSRYLPGISIRNGILSNPGKEFIPPNPWSSSGLSGKSSGTMSVISKVNSVGSS | |
| 25 μL | |
| Protein Kinase | |
| 22858 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction