missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Icilin

missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
Description
Cooling agent that activates the novel cold receptors TRPM8 (CMR1) and TRPA1 (ANKTM1/TRPN1), members of the TRP ion channel family. Induces currents in CMR1-expressing HEK 293 cells (EC50 = 0.36 μM) more potently than menthol or low temperatures. Produces "wet shakes" in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 311.3 |
| Purity | >98% |
| Molecular Weight (g/mol) | 311.297 |
| Chemical Name or Material | Icilin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction