missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICH/USP Parenteral Combined-1 Target Elements: Mo @ 1500, Cr @ 1100, Ba @ 700, Sn @ 600, Cu @ 300, Li @ 250, Sb @ 90, Se @ 80, Ag @ 10, Tl @ 8 μg/mL in 5% HNO3, tr. HF, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
ICH/USP Parenteral Combined-1 Target Elements: Mo @ 1500, Cr @ 1100, Ba @ 700, Sn @ 600, Cu @ 300, Li @ 250, Sb @ 90, Se @ 80, Ag @ 10, Tl @ 8 μg/mL in 5% HNO3, tr. HF
Specifications
Specifications
| Chemical Name or Material | Thallium/Silver/Selenium/Antimony/Lithium/Copper/Tin/Barium/Chromium/Molybdenum |
| CAS | 7440-28-0, 7440-22-4, 7782-49-2, 7440-36-0, 7439-93-2, 7440-50-8 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Thallium/Silver/Selenium/Antimony/Lithium/Copper/Tin/Barium/Chromium/Molybdenum |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Thallium 8 μg/mL/Silver 10 μg/mL/Selenium 80 μg/mL/Antimony 90 μg/mL/Lithium 250 μg/mL/Copper 300 μg/mL/Tin 600 μg/mL/Barium 700 μg/mL/Chromium 1100 μg/mL/Molybdenum 1500 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction