missing translation for 'onlineSavingsMsg'
Learn More

ICH/USP Oral Target Elements Standard D: Cr @ 11,000; Sn @ 6000; Cu, Mo @ 3000; Ba @ 1400; Sb @ 1200; Li @ 550 μg/mL in 5% HNO3/tr. HF, VHG™

Product Code. 17929389
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17929389 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17929389 Supplier VHG Labs Supplier No. VHGICHUSPTELD100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICH/USP Oral Target Elements Standard D: Cr @ 11,000; Sn @ 6000; Cu, Mo @ 3000; Ba @ 1400; Sb @ 1200; Li @ 550 μg/mL in 5% HNO3/tr. HF
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium/Antimony/Barium/Copper/Molybdenum/Tin/Chromium
CAS 7439-93-2, 7440-36-0, 7440-39-3, 7440-50-8, 7439-98-7, 7440-31-5
Methods ICP/ICP-MS
Analyte or Component Names Lithium/Antimony/Barium/Copper/Molybdenum/Tin/Chromium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Lithium 550 μg/mL/Antimony 1200 μg/mL/Barium 1400 μg/mL/Copper 3000 μg/mL/Molybdenum 3000 μg/mL/Tin 6000 μg/mL/Chromium 11000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.