missing translation for 'onlineSavingsMsg'
Learn More

ICH/USP Oral Target Elements Standard B: Ni @ 200; Ag, Se @ 150; V @ 100; Co @ 50; Tl @ 8 μg/mL in 2% HNO3, VHG™

Product Code. 17961076
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17961076 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17961076 Supplier VHG Labs Supplier No. VHGICHUSPTELB100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICH/USP Oral Target Elements Standard B: Ni @ 200; Ag, Se @ 150; V @ 100; Co @ 50; Tl @ 8 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thallium/Cobalt/Vanadium/Selenium/Silver/Nickel
CAS 7440-28-0, 7440-48-4, 7440-62-2, 7782-49-2, 7440-22-4, 7440-02-0
Methods ICP/ICP-MS
Analyte or Component Names Thallium/Cobalt/Vanadium/Selenium/Silver/Nickel
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Thallium 8 μg/mL/Cobalt 50 μg/mL/Vanadium 100 μg/mL/Selenium 150 μg/mL/Silver 150 μg/mL/Nickel 200 μg/mL
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.