missing translation for 'onlineSavingsMsg'
Learn More

ICH/USP Class 1&2A Combined Target Elements: Ni @ 20; As @ 15; V @ 10; Co, Pb @ 5; Hg @ 3; Cd @ 2 μg/mL in 2% HNO3, VHG™

Product Code. 17934386
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17934386 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17934386 Supplier VHG Labs Supplier No. VHGICHUSPTELE100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICH/USP Class 1&2A Combined Target Elements: Ni @ 20; As @ 15; V @ 10; Co, Pb @ 5; Hg @ 3; Cd @ 2 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cadmium/Mercury/Cobalt/Lead/Vanadium/Arsenic/Nickel
CAS 7440-43-9, 7439-97-6, 7440-48-4, 7439-92-1, 7440-62-2, 7440-38-2
Methods ICP/ICP-MS
Analyte or Component Names Cadmium/Mercury/Cobalt/Lead/Vanadium/Arsenic/Nickel
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Cadmium 2 μg/mL/Mercury 3 μg/mL/Cobalt 5 μg/mL/Lead 5 μg/mL/Vanadium 10 μg/mL/Arsenic 15 μg/mL/Nickel 20 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.