Learn More
BINDER™ ICH Light Module
Product Code. 30238258 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
KB PRO 720, KBF/KBF PRO 720
KB PRO/KBF/KBF PRO 260/470
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
30238258 KB PRO/KBF/KBF PRO 260/470 Each
30238384 KB PRO 720, KBF/KBF PRO 720 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30238258 Supplier BINDER™ Supplier No. 80122441
ICH-Q1B light module
Module consists of:
  • VIS cassette
  • UVA cassette
  • Independent power supply
Specifications
For Use With (Equipment) KB PRO/KBF/KBF PRO 260/470
Material Glass
Type Lighting