missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICG-001, MedChemExpress
Description
ICG-001 is an inhibitor of β-catenin/TCF mediated transcription. ICG-001 works by specifically binding to cyclic AMP response element-binding protein with an IC50 of 3 μM. ICG-001 selectively blocks the β-catenin/CBP interaction without interfering with the β-catenin/p300 interaction.
Specifications
Specifications
| Chemical Name or Material | ICG-001 |
| CAS | 780757-88-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C33H32N4O4 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 50 mg/mL (91.14 mM) |
| SMILES | O=C(NCC1=CC=CC=C1)N2CCC(N([C@@]2(CN(C3=O)CC4=C5C=CC=CC5=CC=C4)[H])[C@H]3CC6=CC=C(C=C6)O)=O |
| Molecular Weight (g/mol) | 548.63 |
| Formula Weight | 548.63 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?