missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICEBERG Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06510MFV450
This item is not returnable.
View return policy
Description
ICEBERG Polyclonal antibody specifically detects ICEBERG in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ICEBERG | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| caspase recruitment domain family, member 18, Caspase-1 inhibitor Iceberg, ICEBERG caspase-1 inhibitor, ICEBERGcaspase recruitment domain-containing protein 18, pseudo-ICE, UNQ5804 | |
| Produced in rabbits immunized with E. coli-derived Human ICEBERG fragment. | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 59082 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction