Learn More
Leica™ ICC50 W Wireless HD Camera
Product Code. 15475934
Click to view available options
Camera Type:
Wireless HD
Unit Size:
Each
Product Code. 15475934
Brand: Leica™ 13613735
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Share, Capture and Archive Images with this integrated high definition camera system. ICC50 W Wireless HD Camera provides excellent image sharpness, brightness and color impression.
  • WiFi mode to wirelessly broadcast an HD image directly to your mobile device for independent annotation and image capture by several students
  • Download the Leica AirLab App for camera setup, annotations, measuring, image capture, and sharing to email, photo folders, or other social media connections
  • USB mode to connect directly by cable to your PC for fastest live image of moving objects
  • The Leica ICC50 W is compatible with the full range of Leica Imaging software modules for versatility.
  • Ethernet mode to connect to your own network and allow for the maximum number of mobile devices to access the image
  • SD mode to capture directly onto a memory card when mobile devices and PC's are not available
  • Always connect to an HD display for maximum flexibility
  • Handheld remote for convenient fine tuning of camera settings, capturing onto the SD card, and viewing the gallery of images saved on the SD card
  • The Leica ICC50 W can be powered directly from the Leica DM500 or Leica DM750 stands minimizing infrastructure and cost. Power by external power supply or PC is also possible.
  • Modular design allows for easy upgrades and service
Specifications
Camera Type Wireless HD
Color 24-bit
Height (Metric) 50 mm
Includes Camera, USB cable, HDMI cable, disc with LAS EZ and Leica Acquire software
Weight (Metric) 700 g