missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35534-100ul
This item is not returnable.
View return policy
Description
ICB1 Polyclonal antibody specifically detects ICB1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ICB1 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| basement membrane-induced, chromosome 1 open reading frame 38, ICB1, ICB-1, Induced by contact to basement membrane 1 protein, Protein ICB-1, protein THEMIS2, THEMIS2, Thymocyte-expressed molecule involved in selection protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ICB1 (NP_004839.2).,, Sequence:, MEPVPLQDFVRALDPASLPRVLRVCSGVYFEGSIYEISGNECCLSTGDLIKVTQVRLQKVVCENPKTSQTMELAPNFQGYFTPLNTPQSYETLEELVSATTQSSKQLPTCFMSTHRIVTEGRVVTEDQLLMLEAVVMHLGIRSARCVLGMEGQQVILHLPLSQKGPFWTWEPSAPRTLLQVLQDPALKDLVLTCPTLPWHSLILRPQYEIQAIMHIFSSLRIAATRSAAQTQGEDLARVHQGWLQYVQQDSCPQEGPQAR | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9473 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction