missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICB-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551201
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total C1orf38 / THEMIS2 protein.
May constitute a control point in macrophage inflammatory response, promoting LPS-induced TNF production.
Specifications
| ICB-1 | |
| Polyclonal | |
| Unconjugated | |
| THEMIS2 | |
| basement membrane-induced; basement membrane-induced protein; C1orf38; Icb1; ICB-1; Induced by contact to basement membrane 1 protein; protein ICB-1; Protein THEMIS2; RGD1561016; THEMIS2; thymocyte selection associated family member 2; thymocyte-expressed molecule involved in selection protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9473 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q5TEJ8 | |
| THEMIS2 | |
| Fusion protein of human THEMIS2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction