missing translation for 'onlineSavingsMsg'
Learn More

ICAT/CTNNBIP1 Antibody (536017) [HRP], Novus Biologicals™

Product Code. 30736771 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736771 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736771 Supplier Novus Biologicals Supplier No. FAB3016H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ICAT/CTNNBIP1 Monoclonal antibody specifically detects ICAT/CTNNBIP1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAT/CTNNBIP1
Applications Immunohistochemistry
Classification Monoclonal
Clone 536017
Conjugate HRP
Formulation PBS
Gene Alias beta-catenin-interacting protein ICAT, catenin, beta interacting protein 1, ICATbeta-interacting protein 1, Inhibitor of beta-catenin and Tcf-4
Host Species Mouse
Immunogen E. coli-derived recombinant human ICAT, Met1-Gln81, Accession # Q9NSA3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 56998
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.