missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Icaritin, MedChemExpress
Description
Icaritin (Anhydroicaritin) is a prenylflavonoid derivative from Epimedium Genusis and potently inhibits proliferation of K562 cells (IC50 of 8 μM) and primary CML cells (IC50 of 13.4 μM for CML-CP and 18 μM for CML-BC). Icaritin can regulate MAPK/ERK/JNK and JAK2/STAT3 /AKT signalings, also enhances osteogenesis[3.
Specifications
Specifications
| Chemical Name or Material | Icaritin |
| CAS | 118525-40-9 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H20O6 |
| Quantity | 5 mg |
| Synonym | Anhydroicaritin |
| Solubility Information | DMSO : 15.62 mg/mL (42.40 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C1C(O)=C(C2=CC=C(OC)C=C2)OC3=C(C/C=C(C)\C)C(O)=CC(O)=C13 |
| Molecular Weight (g/mol) | 368.38 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?