missing translation for 'onlineSavingsMsg'
Learn More

Icariside I, MedChemExpress

Product Code. 16451658
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451658 5 mg 5mg
16433936 25 mg 25mg
16409248 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451658 Supplier Medchem Express Supplier No. HYN1939

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Icariside I is a metabolite of Icarlin, which could regulate bone remodeling and is recognized as an effective agent for the treatment of osteoporosis.

Icariside I is a metabolite of Icarlin, which could regulate bone remodeling and is recognized as an effective agent for the treatment of osteoporosis.

Specifications

Chemical Name or Material Icariside I
CAS 56725-99-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H30O11
Quantity 5 mg
Synonym Icarisid I
Solubility Information DMSO : 62.5 mg/mL (117.81 mM; Need ultrasonic)
SMILES O=C1C(O)=C(C2=CC=C(OC)C=C2)OC3=C(C/C=C(C)\C)C(O[C@H]4[C@@H]([C@H]([C@@H]([C@@H](CO)O4)O)O)O)=CC(O)=C13
Molecular Weight (g/mol) 530.52
Formula Weight 530.52
Percent Purity 98.36%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.