missing translation for 'onlineSavingsMsg'
Learn More

ICAP-1 Antibody [DyLight 350], Novus Biologicals™

Product Code. 30738130 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738130 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738130 Supplier Novus Biologicals Supplier No. AF6805D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ICAP-1 Polyclonal antibody specifically detects ICAP-1 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAP-1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias bodenin, DKFZp686K08158, ICAP-1, ICAP-1alpha, ICAP-1B, ICAP1ICAP1AICAP1BICAP-1A, integrin beta 1 binding protein 1, integrin beta-1-binding protein 1, Integrin cytoplasmic domain-associated protein 1, integrin cytoplasmic domain-associated protein 1-alpha, integrin cytoplasmic domain-associated protein 1-beta
Host Species Sheep
Immunogen E. coli-derived recombinant human ICAP-1, Met1-Tyr127, Accession # O14713
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9270
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.