missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17699-1-AP-20UL
This item is not returnable.
View return policy
Description
SOS2 promotes the exchange of Ras-bound GDP by GTP.Specifications
| ICAM3 | |
| Polyclonal | |
| Unconjugated | |
| ICAM3 | |
| CD50, CDW50, ICAM 3, ICAM R, ICAM3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3385 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Flow Cytometry | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P32942 | |
| ICAM3 | |
| ICAM3 Fusion Protein Ag11955 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction