missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ICAM3 Monoclonal Antibody (76205)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561915

Product Code. 30535557

  • £490.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

CD50 (intracellular adhesion molecule 3, ICAM-3) is a transmembrane glycoprotein expressed by leukocytes, that serves as a counter-receptor for the lymphocyte function-associated antigen (LFA)-1 integrin. Besides functioning as an adhesive molecule that mediates e.g. the contact between T cells and antigen presenting cells, ICAM-3 regulates affinity of LFA-1 for ICAM-1 and induces T cell activation and proliferation. ICAM-3 plays an essential role in the initiation of the immune response both on T cells and antigen presenting cells and interacts also with CD209 (dendritic cell-specific ICAM-3-grabbing nonintegrin, DC-SIGN), a C-type lectin of dendritic cells and macrophages; this process is involved in dialogue between dendritic cells and granulocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ICAM3
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P32942
ICAM3
Mouse myeloma cell line NS0-derived recombinant human ICAM-3/CD50, Gln30-His485.
500 μg
Primary
Human
Antibody
IgG1
Inhibition Assays, Western Blot
76205
Unconjugated
ICAM3
CD50; CDw50; ICAM3; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3
Mouse
Protein A/G
RUO
3385
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.