missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10121-2-AP-150UL
This item is not returnable.
View return policy
Description
This antibody got 55-80 kDa in western blotting maybe due to glycosylation.
ICAM proteins are ligands for the leukocyte adhesion protein LFA-1 (integrin alpha-L/beta-2). ICAM2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance.Specifications
| ICAM2 | |
| Polyclonal | |
| Unconjugated | |
| ICAM2 | |
| CD102, ICAM 2, ICAM2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15896, 3384, 360647 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P13598, P35330 | |
| ICAM2 | |
| ICAM2 Fusion Protein Ag0165 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction