missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27322-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody got 55-80 kDa in western blotting maybe due to glycosylation.
The protein encoded by this gene contains an N-terminal forkhead domain, two BRCA1 C-terminal (BRCT) motifs and a central domain with 13 repetitions of an approximately 41-amino acid sequence. The encoded protein is required to activate the intra-S phase and G2/M phase cell cycle checkpoints in response to DNA damage. This nuclear protein interacts with phosphorylated histone H2AX near sites of DNA double-strand breaks through its BRCT motifs, and facilitates recruitment of the ATM kinase and meiotic recombination 11 protein complex to DNA damage foci.Specifications
| ICAM2 | |
| Polyclonal | |
| Unconjugated | |
| ICAM2 | |
| CD102, ICAM 2, ICAM2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15896, 3384 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P13598, P35330 | |
| ICAM2 | |
| ICAM2 Fusion Protein Ag25952 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction