missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICAM-5 Monoclonal Antibody (191321)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA560932
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors. Appears to be involved in the formation of the initiation complex at the promoter by mediating the interaction between Pol I and UBTF/UBF.
Specifications
| ICAM-5 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q60625 | |
| Icam5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ICAM-5, Leu31-Arg828 (Pro47Arg). | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 191321 | |
| Unconjugated | |
| Icam5 | |
| CD50; Icam3; ICAM-3; Icam5; ICAM-5; intercellular adhesion molecule 3; intercellular adhesion molecule 5; intercellular adhesion molecule 5, telencephalin; telencephalin; Tlcn; TLN | |
| Rat | |
| Protein A/G | |
| RUO | |
| 15898 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction