missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICAM-4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112917
This item is not returnable.
View return policy
Description
ICAM-4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes the Landsteiner-Wiener (LW) blood group antigen(s) that belongs to the immunoglobulin (Ig) superfamily, and that shares similarity with the intercellular adhesion molecule (ICAM) protein family. This ICAM protein contains 2 Ig-like C2-type domains and binds to the leukocyte adhesion LFA-1 protein. The molecular basis of the LW(A)/LW(B) blood group antigens is a single aa variation at position 100; Gln-100=LW(A) and Arg-100=LW(B). Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
| ICAM-4 | |
| Polyclonal | |
| Unconjugated | |
| ICAM4 | |
| 1810015M19Rik; CD242; CD242 antigen; ICAM4; ICAM-4; Intercellular adhesion molecule 4; intercellular adhesion molecule 4 (Landsteiner-Wiener blood group); intercellular adhesion molecule 4 (LW blood group); intercellular adhesion molecule 4 isoform 3 anitgen LWa; intercellular adhesion molecule 4, Landsteiner-Wiener blood group; Landsteiner-Wiener blood group antigen a; Landsteiner-Wiener blood group glycoprotein; LW; LW blood group protein; LW glycoprotein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3386, 78369 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14773, Q9ERM2 | |
| ICAM4 | |
| Recombinant Human Intercellular adhesion molecule 4 protein (20-140AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction