Learn More
ICAM-4 (CD242) Mouse anti-Human, Brilliant Violet 480, Clone: 729632, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 748053
This item is not returnable.
View return policy
Description
ICAM-4 (CD242) Monoclonal antibody specifically detects ICAM-4 (CD242) in Human samples. It is validated for Flow Cytometry.
Specifications
| ICAM-4 (CD242) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| ICAM-4 (CD242) | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 729632 | |
| Brilliant Violet 480 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG2b κ |