missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-4 Antibody (729632), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8397CL1
This item is not returnable.
View return policy
Description
ICAM-4 Monoclonal antibody specifically detects ICAM-4 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| ICAM-4 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD242, CD242 antigen, ICAM-4, intercellular adhesion molecule 4, intercellular adhesion molecule 4 (Landsteiner-Wiener blood group), intercellular adhesion molecule 4 (LW blood group), intercellular adhesion molecule 4, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group antigen a, Landsteiner-Wiener blood group glycoprotein, LWLW blood group protein | |
| Chinese hamster ovary cell line CHO-derived recombinant human ICAM-4, Ala23-Ala240, Accession # Q14773 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 729632 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3386 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction