missing translation for 'onlineSavingsMsg'
Learn More

ICAM-3/CD50 Rabbit anti-Human, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody

£286.00 - £429.00

Specifications

Antigen ICAM-3/CD50
Clone 2
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18201178
View Documents
Novus Biologicals
NBP2-89426-50UL
50 μg
£286.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18216997
View Documents
Novus Biologicals
NBP2-89426-100UL
100 μg
£429.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ICAM-3/CD50
Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Monoclonal
Purified
RUO
Human
CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3
Recombinant Human ICAM-3/CD50 protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
2
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cellular Markers, Immunology
0.2 um filtered solution in PBS
3385
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.