missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-3/CD50 Antibody (ICAM3/1019) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48227
This item is not returnable.
View return policy
Description
ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ICAM-3/CD50 | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| ICAM3 | |
| Recombinant human ICAM-3/CD50 protein (Uniprot: P32942) | |
| 7 mL | |
| Cellular Markers, Immunology | |
| 3385 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| ICAM3/1019 | |
| Immunohistochemistry-Paraffin | |
| CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes an N-glycosylated glycoprotein of 120kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3. CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This monoclonal antibody is excellent for staining of formalin/paraffin tissues. | |
| Store at 4C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction