missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2/CD102 Rabbit anti-Human, Clone: 55, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89425-100UL
This item is not returnable.
View return policy
Description
ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot.
Specifications
| ICAM-2/CD102 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 55 | |
| Western Blot 1:500-1:1000 | |
| CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
| Recombinant Human ICAM-2/CD102 protein | |
| 100 μg | |
| Adaptive Immunity, Immunology | |
| 3384 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction