missing translation for 'onlineSavingsMsg'
Learn More

ICAM-2/CD102 Mouse anti-Human, Clone: B-T1, Novus Biologicals™

Product Code. 18356963 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356963 200 μg 200µL
18383715 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18356963 Supplier Novus Biologicals Supplier No. NBP314603200UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ICAM-2/CD102 Monoclonal antibody specifically detects ICAM-2/CD102 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAM-2/CD102
Applications Flow Cytometry, ELISA, Immunoprecipitation, Functional Assay
Classification Monoclonal
Clone B-T1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA, Immunoprecipitation, Functional
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2
Host Species Mouse
Immunogen ICAM-2 transfected CHO cell line
Purification Method Affinity purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3384
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.