missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2/CD102 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32236AF488
This item is not returnable.
View return policy
Description
ICAM-2/CD102 Polyclonal antibody specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot
Specifications
| ICAM-2/CD102 | |
| Polyclonal | |
| Western Blot | |
| Immunogen affinity purified | |
| Adaptive Immunity, Immunology | |
| Store at 4°C in the dark. |
| Western Blot | |
| Alexa Fluor 488 | |
| CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
| RUO | |
| Primary | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction