missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2/CD102 Antibody (033), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89423AF647
This item is not returnable.
View return policy
Description
ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot.
Specifications
| ICAM-2/CD102 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ICAM-2/CD102 (NP_000864.2; Met1-Gln223). | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3384 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 33 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction