Learn More
ICAM-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15364-1-AP-20UL
 View more versions of this product
Product Code. 16894514
  • £135.00 / 20µL
Description
Intercellular adhesion molecule-1 (ICAM-1, CD54) is an integral membrane protein, a member of the immunoglobulin superfamily, and a ligand for lymphocyte function-associated (LFA) antigens, a beta 2 leukocyte integrin. The normal function of human ICAM-1 is to provide adhesion between endothelial cells and leukocytes after injury or stress. ICAM-1 binds to leukocyte function-associated antigen (LFA-1) or macrophage-1 antigen (Mac-1). It is found on leukocytes, fibroblasts, epithelial cells and endothelial cells and its expression is regulated by inflammatory cytokines. ICAM-1 has a tissue distribution similar to that of the major histocompatibility complex class II antigens and is likely to play a role in inflammatory responses.
Specifications
Specifications
ICAM-1
Polyclonal
Unconjugated
ICAM1
BB2, CD54, ICAM 1, ICAM1, P3.58
Rabbit
Antigen Affinity Chromatography
RUO
3383
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Flow Cytometry
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P05362
ICAM1
ICAM-1 Fusion Protein Ag8309
20 μL