missing translation for 'onlineSavingsMsg'
Learn More

ICAM-1/CD54 Antibody (MEM-111) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications

£149.00 - £413.00

Specifications

Antigen ICAM-1/CD54
Clone MEM-111
Concentration 1 mg/mL
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18757353
View Documents
Novus Biologicals
NB500-318
0.1 mg
£413.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18501231
View Documents
Novus Biologicals
NB500-318-25ug
25 μg
£149.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications

Specifications

ICAM-1/CD54
1 mg/mL
Monoclonal
Purified
RUO
Human, Rat, Bovine
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
ICAM1
IgG2a
Protein A purified
MEM-111
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Mouse
Adaptive Immunity, Cancer, Cell Biology, Immunology
P05362
3383
Raji human Burkitt's lymphoma cell line (NM_000201.2).
Primary
110 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.