missing translation for 'onlineSavingsMsg'
Learn More

ICAM-1/CD54 Antibody (3E2B), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18403327 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403327 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403327 Supplier Novus Biologicals Supplier No. NBP222540MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hamster Monoclonal Antibody

ICAM-1/CD54 Monoclonal antibody specifically detects ICAM-1/CD54 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Inhibition Activity, CyTOF, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAM-1/CD54
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition Assays, Blocking Assay, Blocking Assay
Classification Monoclonal
Clone 3E2B
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Inhibition Activity, CyTOF-ready, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Host Species Hamster
Immunogen Mouse ICAM-1 (CD54)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3383
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.