Learn More
Invitrogen™ ICAD Recombinant Rabbit Monoclonal Antibody (24GB4810)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557380
 View more versions of this product
Product Code. 30276060
  • £172.00 / 20µL
Description
ICAD Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
ICAD is the inhibitor of caspase-3-activated DNase which is a substrate that controls nuclear apoptosis. ICAD has two isoforms: a functional isoform of 45kDa, ICAD-L/DNA fragmentation factor (DFF) 45; and a 35kDa isoform, ICAD-S/DFF35. Although both ICAD-L and ICAD-S can bind and inhibit CAD, only ICAD-L was reported to be functional. ICAD is cleaved to be inactivated and allow caspase-activated DNase (CAD) to execute nuclear internucleosomal apoptotic DNA fragmentation. In non-apoptotic cells, CAD is complexed with its inhibitor, ICAD. The activation of the CAD/ICAD complex occurs through the caspase 3-mediated cleavage of ICAD at residues 117 and 224, which results in three ICAD fragments that are then released from CAD. In addition to its DNase inhibitory activity, ICAD acts as a CAD specific folding chaperone. There are recent reports that ICAD is a potential target for restoring a normal apoptotic signal transduction pathway in colon and brain cancer cells. Two alternatively spliced transcript variants encoding distinct isoforms have been found for the ICAD gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ICAD
Recombinant Monoclonal
Unconjugated
DFFA
A330085O09Rik; DFF; DFF1; DFF35; Dff45; DFF-45; Dffa; DNA fragmentation factor 45 kDa subunit; DNA fragmentation factor subunit alpha; DNA fragmentation factor, 45kDa, alpha polypeptide; DNA fragmentation factor, alpha subunit; H13; ICAD; ICAD-L; ICAD-S; Inhibitor of CAD; RP23-121D17.3
Rabbit
Affinity chromatography
RUO
1676
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4810
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O00273
DFFA
A synthesized peptide derived from human ICAD (aa 318-331).
20 μL