missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICAD (cleaved Asp224) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577941
This item is not returnable.
View return policy
Description
Positive Control: Jurkat, Jurkat (10uM etoposide treatment for 24h) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Apoptosis is a cell death process that removes toxic and/or useless cells during mammalian development. The apoptotic process is accompanied by shrinkage and fragmentation of the cells and nuclei and degradation of the chromosomal DNA into nucleosomal units. DNA fragmentation factor (DFF) is a heterodimeric protein of 40-kD (DFFB) and 45-kD (DFFA) subunits. DFFA is the substrate for caspase-3 and triggers DNA fragmentation during apoptosis. DFF becomes activated when DFFA is cleaved by caspase-3. The cleaved fragments of DFFA dissociate from DFFB, the active component of DFF. DFFB has been found to trigger both DNA fragmentation and chromatin condensation during apoptosis. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| ICAD (cleaved Asp224) | |
| Polyclonal | |
| Unconjugated | |
| DFFA | |
| DFF1; DFF45; DFF-45; DFFA; DNA fragmentation factor 45 kDa subunit; DNA fragmentation factor subunit alpha; DNA fragmentation factor, 45kDa, alpha polypeptide; H13; ICAD; Inhibitor of CAD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1676 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.73 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O00273 | |
| DFFA | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ICAD (cleaved Asp224). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction