missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ICA 121431

Product Code. 30002058
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002058 50 mg 50mg
30002050 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002058 Supplier Tocris Bioscience™ Supplier No. 5066/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective NaV1.3 and NaV1.1 channel blocker

Potent and selective inhibitor of human NaV1.3 and NaV1.1 channels (IC50 values are 13 and 23 nM respectively). Exhibits up to 1,000 fold selectivity against other TTX-sensitive or resistant sodium channels (IC50 values are >10 μM for human NaV1.5 and NaV1.8 channels). Interacts with an inhibitory interaction site distinct from those bound by TTX and local anesthetic-like modulators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313254-51-2
Molecular Formula C23H19N3O3S2

Specifications

Quantity 50 mg
Formula Weight 449.55
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 449.543
Chemical Name or Material ICA 121431
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.