missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICA-121431, MedChemExpress
Description
ICA-121431 is a nanomolar potent and broad-spectrum voltage-gated sodium channel (Nav) blocker, shows equipotent selectivity for human Nav1.1 and Nav1.3 subtypes with IC50 values of 13 nM and 23 nM, respectively. ICA-121431 shows less potent inhibition of Nav1.2 (IC50=240 nM) and 1,000 fold selectivity against Nav1.4, Nav1.6, and the TTX-resistant human Nav1.5 and Nav1.8 channels (IC50s >10 μM).
Specifications
Specifications
| Chemical Name or Material | ICA-121431 |
| CAS | 313254-51-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H19N3O3S2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 44 mg/mL (97.88 mM) |
| SMILES | O=C(NC1=CC=C(S(NC2=NC=CS2)(=O)=O)C=C1)C(C3=CC=CC=C3)C4=CC=CC=C4 |
| Molecular Weight (g/mol) | 449.55 |
| Formula Weight | 449.55 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?