missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ICA 069673
GREENER_CHOICE

Product Code. 18717046
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18717046 25 mg 25mg
18795356 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18717046 Supplier Tocris Bioscience™ Supplier No. 4305/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KV7.2/KV7.3 channel opener

ICA 069673 is a selective KV7.2/KV7.3 (KCNQ2/3) channel opener (EC50 = 0.69 μM). Exhibits 20-fold selectivity for KV7.2/KV7.3 over KV7.3/KV7.5, with no measurable activity over a panel of cardiac ion channels. Hyperpolarizes resting membrane potential and inhibits spontaneous action potentials in guinea pig detrusor muscle (DSM) isolated cells. Orally active in animal models of epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Chloro-5-pyrimidinyl)-3,4-difluorobenzamide
CAS 582323-16-8
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C11H6ClF2N3O
Formula Weight Observed MW: 269.63
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.