missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ IC-XT Pack Sanitizer Neutralizing Agar, RODAC™ LL Plates
Click to view available options
:
IC-XT Sanitizer Neutralizing Agar: RODAC LL Plates x 100
Description
IC-XT (Isolator Cleanroom-Extended Temperature) Pack products are gamma-sterilized to allow monitoring of environmental and product hygiene in clean rooms of pharmaceutical production and fill rooms, and in isolators.
- Used for monitoring the efficiency of disinfection in controlled environments
- For use in replicate organism detection and counting procedures
- Medium specified in the United States Pharmacopeia and European Pharmacopeia for the total aerobic microbial count testing procedures
- For surface testing, apply the plate surface directly to the surface being tested and exert moderate pressure (without rubbing or moving the agar surface
- Areas (walls, floors etc.) may be divided into sections or grids and samples taken from specific points within the grid.
RODAC™ (Replicate Organism Detection And Counting) Locking Lid Media Plates
- Particularly recommended for monitoring surfaces of sanitary importance
- Enables convenient handling while reducing the risk of accidental contamination
- Locking Lid (LL) mechanism allows for easy locking of plates for safe transport
- Ensures a secure fit between lid and base to prevent unintentional opening of plates
- Locked plates allow for appropriate aeration during incubation
- Plates are provided in the unlocked position for easy removal of lid
- Simply twist the base and the lid into the desired position to lock or unlock plate
- Packaged in impermeable plastic films to allow an extended stability and storage
Specifications
Specifications
| Cap Type | Locking Lid |
| Certifications/Compliance | USP, EP, ISO 11137 |
| Content And Storage | 2°-25° C (ideal: 15°-22° C) |
| Detects | Microorganisms |
| Format | Prepared BODAC plates |
| Product Type | Agar |
| Sterility | Sterile |
| Formulation | Per 1 L: Pancreatic Digest of Casein 15 g, Papaic Digest of Soybean 5 g, Sodium Chloride 5 g, Lecithin 0.7 g, Polysorbate 80 5 g, Sodium Thiosulphate 0.5 g, Sodium Pyruvate 3.5 g, L-Histidine 1 g, Agar 18.5 g |
| For Use With (Application) | Detection of microorganisms surviving after disinfection treatment |
| Includes | SORB-IT™ silica gel desiccant bag |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction