missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibutamoren Mesylate, MedChemExpress
Description
Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. Ibutamoren Mesylate is an orally active growth hormone (GH) secretagogue.
Specifications
Specifications
| Chemical Name or Material | Ibutamoren Mesylate |
| CAS | 159752-10-0 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C28H40N4O8S2 |
| Quantity | 10 mg |
| Synonym | MK-677 MK-0677 |
| Solubility Information | DMSO : 50 mg/mL (80.03 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (80.03 mM) |
| SMILES | CC(N)(C(N[C@@H](C(N1CCC2(CN(C3=C2C=CC=C3)S(=O)(C)=O)CC1)=O)COCC4=CC=CC=C4)=O)C.CS(=O)(O)=O |
| Molecular Weight (g/mol) | 624.77 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?