missing translation for 'onlineSavingsMsg'
Learn More

Ibutamoren Mesylate, MedChemExpress

Product Code. 16431926
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431926 500 mg 500mg
16458108 10 mg 10mg
16450568 5 mg 5mg
16434027 50 mg 50mg
16417876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16431926 Supplier Medchem Express Supplier No. HY50844

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. Ibutamoren Mesylate is an orally active growth hormone (GH) secretagogue.

Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. Ibutamoren Mesylate is an orally active growth hormone (GH) secretagogue.

Specifications

Chemical Name or Material Ibutamoren Mesylate
CAS 159752-10-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H40N4O8S2
Quantity 500 mg
Synonym MK-677 MK-0677
Solubility Information DMSO : 50 mg/mL (80.03 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (80.03 mM)
SMILES CC(N)(C(N[C@@H](C(N1CCC2(CN(C3=C2C=CC=C3)S(=O)(C)=O)CC1)=O)COCC4=CC=CC=C4)=O)C.CS(=O)(O)=O
Molecular Weight (g/mol) 624.77
Formula Weight 624.77
Percent Purity 97.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.