missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen Sodium Salt, TRC

Product Code. 30407199
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407199 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407199 Supplier Toronto Research Chemicals Supplier No. TRCI400158100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ibuprofen Sodium Salt
CAS 31121-93-4
Purity Grade Notes HPLC
Molecular Formula C13H17NaO2
Quantity 100 mg
Synonym Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, sodium salt (1:1) (ACI), Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, sodium salt (9CI), Hydratropic acid, p-isobutyl-, sodium salt (8CI), 2-(4-Isobutylphenyl)propionic acid sodium salt, Ibuprofen sodium, Ibuprofen sodium salt, Sodium 2-(4-isobutylphenyl)propionate, Sodium ibuprofen, α-Methyl-4-(2-methylpropyl)benzeneacetic acid sodium salt
InChI Formula InChI=1S/C13H18O2.Na/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15;/h4-7,9-10H,8H2,1-3H3,(H,14,15);/q;+1/p-1
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(=O)[O-].[Na+]
IUPAC Name sodium;2-[4-(2-methylpropyl)phenyl]propanoate
Molecular Weight (g/mol) 228.26
Formula Weight 228.1126
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.