missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen Piconol, TRC

Product Code. 30391399
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30391399

Brand: Toronto Research Chemicals TRCI4001501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ibuprofen Piconol
CAS 64622-45-3
Purity Grade Notes HPLC
Molecular Formula C19 H23 N O2
Quantity 1 g
Synonym Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, 2-pyridinylmethyl ester, 2-pyridylmethyl 2-(4-isobutylphenyl)propanoate
InChI Formula InChI=1S/C19H23NO2/c1-14(2)12-16-7-9-17(10-8-16)15(3)19(21)22-13-18-6-4-5-11-20-18/h4-11,14-15H,12-13H2,1-3H3
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)OCc2ccccn2
IUPAC Name pyridin-2-ylmethyl 2-[4-(2-methylpropyl)phenyl]propanoate
Molecular Weight (g/mol) 297.39
Formula Weight 297.1729
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.