missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibuprofen Piconol, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | Ibuprofen Piconol |
| CAS | 64622-45-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H23 N O2 |
| Quantity | 2.5 g |
| Synonym | Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, 2-pyridinylmethyl ester, 2-pyridylmethyl 2-(4-isobutylphenyl)propanoate |
| InChI Formula | InChI=1S/C19H23NO2/c1-14(2)12-16-7-9-17(10-8-16)15(3)19(21)22-13-18-6-4-5-11-20-18/h4-11,14-15H,12-13H2,1-3H3 |
| SMILES | CC(C)Cc1ccc(cc1)C(C)C(=O)OCc2ccccn2 |
| IUPAC Name | pyridin-2-ylmethyl 2-[4-(2-methylpropyl)phenyl]propanoate |
| Molecular Weight (g/mol) | 297.39 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction