missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen, MedChemExpress

Product Code. 16430678
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16430678

Brand: Medchem Express HY78131

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ibuprofen ((±)-Ibuprofen) is a potent, orally active, selective COX-1 inhibitor with an IC50 value of 13 μM. Ibuprofen inhibits cell proliferation, angiogenesis, and induces cell apoptosis. Ibuprofen is a nonsteroidal anti-inflammatory agent and a nitric oxide (NO) donor. Ibuprofen ((±)-Ibuprofen) can be used in the research of pain, swelling, inflammation, infection, immunology, cancers.

Ibuprofen ((±)-Ibuprofen) is a potent, orally active, selective COX-1 inhibitor with an IC50 value of 13 μM. Ibuprofen inhibits cell proliferation, angiogenesis, and induces cell apoptosis. Ibuprofen is a nonsteroidal anti-inflammatory agent and a nitric oxide (NO) donor. Ibuprofen ((±)-Ibuprofen) can be used in the research of pain, swelling, inflammation, infection, immunology, cancers.

Specifications

Chemical Name or Material Ibuprofen
Physical Form Powder
CAS 15687-27-1
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C13H18O2
For Use With (Application) COVID-19-immunoregulation
Synonym (±)-Ibuprofen
Solubility Information DMSO : 100 mg/mL (484.78 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.85 mM; Need ultrasonic)
SMILES OC(C(C1=CC=C(CC(C)C)C=C1)C)=O
Molecular Weight (g/mol) 206.28
Formula Weight 206.28
Percent Purity 99.5%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.