missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen, LoGiCal

Product Code. 30463133
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463133 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463133 Supplier LoGiCal Supplier No. LGCFOR0002.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ibuprofen
CAS 15687-27-1
Analyte or Component Names Ibuprofen
Shipping Condition Room Temperature
Molecular Formula C13 H18 O2
Certifications/Compliance ISO 9001
Synonym alpha-Methyl-4-(2-methylpropyl)-benzeneacetic acid, p-Isobutylhydratropic acid, (+/-)-alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid, (+/-)-2-(p-Isobutylphenyl)propionic acid, (+/-)-Ibuprofen, (+/-)-Ibuprophen, (4-Isobutylphenyl)-alpha-methyl-acetic acid, (RS)-Ibuprofen, alpha-(4-Isobutylphenyl)propionic acid, alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid, 2-(4'-Isobutylphenyl)propionic acid, 2-(4-Isobutylphenyl)propanoic acid, 2-(p-Isobutylphenyl)propionic acid, 4-Isobutylhydratropic acid, Act 3, Actiprofen, Adex 200, Adran, Advil, Alaxan, Algi-Flanderil, Algiflex, Algofen, Am-Fam 400, Amibufen, Anafen, Anco, Andran, Anflagen, Antarene, Antiflam, Apo-Ibuprofen, Apsifen, Artofen, Artril, Artril 300, Atril 300, Balkaprofen, Betaprofen, Bloom, Bluton, Brofen, Brufanic, Brufen, Brufen 400, Brufen Retard, Bruflam, Brufort, Buburone, Buluofen, Burana, Butacortelone, Butylenin, Carol, Cobo, Codral Period Pain, Combiflam, Dansida, Dentigoa, Dibufen, Dolgin, Dolgirid, Dolgit, Dolmaral, Dolo-Dolgit, Dolocyl, Dolofen, Dolofen F, Dolomax, Donjust B, Doretrim, Dorival, Drin, Easifon, Ebufac, Emflam, Emflam 200, Emodin, Epobron, Femadon, Fenbid, Fenspan, Focus, Gofen, Gynofug, Haltran, IB 100, IP 82, Ibosure, Ibren, Ibu-Attritin, Ibu-Tab, Ibu-slow, Ibufen, Ibuflamar, Ibufug, Ibugen, Ibugesic, Ibuleve, Ibulgan, Ibumetin, Ibupirac, Ibupril, Ibuprocin, Ibuprofen, Ibuprohm, Ibusal, Ibutad, Ifen, Inabrin, Inflam, Inza, Ipren, Iprogel, Irfen, Isodol, Lamidon, Librofem, Lidifen, Liptan, Lopane, Mensoton, Motrin, Motrin IB, Mynosedin, NSC 256857, Nagifen-D, Napacetin, Nobafon, Nobfelon, Nobgen, Noritis, Norton, Novogent, Novoprofen, Nuprin, Nurofen, Optifen, Opturem, Ostarin, Ostofen, Paduden, Panafen, Pantrop, Paxofen, Pediaprofen, Perofen, Proartinal, Profen, Proflex, Prontalgin, Provon, Quadrax, RD 13621, Rafen, Ranofen, Recidol, Relcofen, Roidenin, Rufin, Rupan, Seclodin, Stelar, Suspren, Syntofene, Tabalon, Tabalon 400, Tarein, Tatanal, Tofen, Trendar, U 18753, Unipron, Upfen, Uprofen, Urem, Zafen, Zofen, dl-Ibuprofen, p-Isobutyl-2-phenylpropionic acid, 4-Isobutyl-a-methylphenylacetic acid
InChI Formula InChI=1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)
Solution Type Neat
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)O
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 206.28
Quantity 10 mg
Formula Weight 206.1307 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.